Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098101.3 | XP_001098101.2 | 1021 | 62 | ||
| XM_015123703.2 | XP_014979189.1 | 895 | 62 | |||
| XM_028839240.1 | XP_028695073.1 | 747 | 62 | |||
| GenBank mRNA | JU335469.1 | - | 777 | 62 | ||
| JU472331.1 | - | 781 | 62 | |||
| JV045540.1 | - | 675 | 62 | |||
| JV656077.1 | - | 784 | 62 | |||
More Information
Set Membership: |
|

LIN37
Genomic Map