Assay Details
| Gene Symbol: ZNF329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125039.2 | XP_014980525.2 | 164 | 94 | ||
| XM_028840315.1 | XP_028696148.1 | 233 | 94 | |||
| XM_028840316.1 | XP_028696149.1 | 237 | 94 | |||
| GenBank mRNA | JU336403.1 | - | 105 | 94 | ||
| JV723434.1 | - | 83 | 94 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015125039.2, XM_028840315.1, XM_028840316.1 transcripts.
Set Membership: |
|

ZNF329
Genomic Map