Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101623.4 | XP_001101623.2 | 498 | 70 | ||
| GenBank mRNA | JU479807.1 | - | 166 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF13C| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101623.4 | XP_001101623.2 | 498 | 70 | ||
| GenBank mRNA | JU479807.1 | - | 166 | 70 | ||
Set Membership: |
|