Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113899.2 | XP_014969385.1 | 2975 | 94 | ||
| GenBank mRNA | JV463533.1 | - | 326 | 94 | ||
| JV679256.1 | - | 314 | 94 | |||
More Information
Set Membership: |
|

CFAP126
Genomic Map