Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089245.4 | XP_001089245.1 | 1678 | 61 | ||
| GenBank mRNA | JV626416.1 | - | 1684 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BMP7| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089245.4 | XP_001089245.1 | 1678 | 61 | ||
| GenBank mRNA | JV626416.1 | - | 1684 | 61 | ||
Set Membership: |
|