Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135772.2 | XP_014991258.1 | 512 | 69 | ||
| XM_028847762.1 | XP_028703595.1 | 446 | 69 | |||
| GenBank mRNA | JU328483.1 | - | 125 | 69 | ||
| JV521425.1 | - | 377 | 69 | |||
More Information
Set Membership: |
|

ZSCAN16
Genomic Map