Assay Details
Gene Symbol: TRIM39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135874.2 | XP_014991360.1 | 1044 | 97 | ||
XM_015135875.2 | XP_014991361.1 | 897 | 97 | |||
XM_015135876.2 | XP_014991362.1 | 1059 | 97 | |||
XM_015135878.2 | XP_014991364.1 | 1150 | 97 | |||
XM_015135879.2 | XP_014991365.1 | 1039 | 97 | |||
XM_028846903.1 | XP_028702736.1 | 1170 | 97 | |||
GenBank mRNA | JU324789.1 | - | 743 | 97 | ||
JV045345.1 | - | 694 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
