Assay Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802829.3 | XP_002802875.2 | 688 | 115 | ||
| XM_015131219.2 | XP_014986705.1 | 1067 | 115 | |||
| XM_015131221.2 | XP_014986707.1 | 337 | 115 | |||
| XM_015131222.2 | XP_014986708.1 | 695 | 115 | |||
| XM_015131223.2 | XP_014986709.1 | 1064 | 115 | |||
| XM_028844003.1 | XP_028699836.1 | 691 | 115 | |||
| XM_028844004.1 | XP_028699837.1 | 945 | 115 | |||
| GenBank mRNA | JV651068.1 | - | 154 | 115 | ||
More Information
Set Membership: |
|

ZNF662
Genomic Map