Assay Details
| Gene Symbol: LAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128561.2 | XP_014984047.1 | 664 | 119 | ||
| GenBank mRNA | JU332328.1 | - | 331 | 119 | ||
| JU472911.1 | - | 317 | 119 | |||
| JV044963.1 | - | 400 | 119 | |||
More Information
Set Membership: |
|

LAGE3
Genomic Map