Assay Details
| Gene Symbol: HSPA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136003.2 | XP_014991489.2 | 1803 | 98 | ||
| GenBank mRNA | AY680559.1 | - | 121 | 98 | ||
| JU475405.1 | - | 1352 | 98 | |||
| JV489416.1 | - | 1429 | 98 | |||
| Gene Symbol: LOC114669856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847714.1 | XP_028703547.1 | 1552 | 98 | ||
| GenBank mRNA | JU342693.1 | - | 1228 | 98 | ||
| JU475406.1 | - | 1352 | 98 | |||
| JV649991.1 | - | 1482 | 98 | |||
More Information
Set Membership: |
|

HSPA1A
Genomic Map