Assay Details
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838748.1 | XP_028694581.1 | 346 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028838748.1 transcripts.
Set Membership: |
|

IL11
Genomic Map