Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124687.2 | XP_014980173.1 | 679 | 83 | ||
| XM_028839471.1 | XP_028695304.1 | 458 | 83 | |||
| XM_028839472.1 | XP_028695305.1 | 221 | 83 | |||
More Information
Set Membership: |
|

KLK13
Genomic Map