Assay Details
| Gene Symbol: SUCNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100627.4 | XP_001100627.2 | 151 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001100627.4 transcripts.
Set Membership: |
|

SUCNR1
Genomic Map