Assay Details
Gene Symbol: SHOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842081.1 | XP_028697914.1 | 408 | 88 |
Gene Symbol: LOC719166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843006.1 | XP_028698839.1 | 408 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
