Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261007.1 | NP_001247936.1 | 194 | 58 | ||
| GenBank mRNA | JU322820.1 | - | 202 | 58 | ||
| JU474273.1 | - | 194 | 58 | |||
| JV605565.1 | - | 207 | 58 | |||
More Information
Set Membership: |
|

ISCA2
Genomic Map