Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204120.1 | NP_001191049.1 | 96 | 102 | ||
| XM_028834345.1 | XP_028690178.1 | 106 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001204120.1, XM_028834345.1 transcripts.
Set Membership: |
|

PALM2AKAP2
Genomic Map