Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204120.1 | NP_001191049.1 | 2696 | 82 | ||
| NM_001371502.1 | NP_001358431.1 | 3247 | 82 | |||
| XM_028834344.1 | XP_028690177.1 | 2843 | 82 | |||
| GenBank mRNA | JU334592.1 | - | 3247 | 82 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map