Assay Details
| Gene Symbol: Sypl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108563.1 | NP_001102033.1 | 1 - 2 | 236 | 107 | |
| GenBank mRNA | AB158471.1 | - | 1 - 2 | 54 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sypl2
Genomic Map