Assay Details
| Gene Symbol: Skor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243246.2 | XP_006243308.2 | 7 - 8 | 2776 | 67 | |
| XM_008766381.1 | XP_008764603.1 | 7 - 8 | 2655 | 67 | ||
More Information
Set Membership: |
|

Skor1
Genomic Map