Assay Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751632.3 | XP_003751680.1 | 1 - 2 | 1083 | 68 | |
| XM_573950.6 | XP_573950.4 | 1 - 2 | 1083 | 68 | ||
More Information
Set Membership: |
|

Fam155a
Genomic Map