Assay Details
| Gene Symbol: Chrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012527.2 | NP_036659.2 | 5 | 2363 | 129 | |
| XM_008771690.1 | XP_008769912.1 | 6 | 2309 | 129 | ||
| GenBank mRNA | M16407.1 | - | 2 | 1856 | 129 | |
| M18088.1 | - | 3 | 1947 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of vascular smooth muscle contraction
smooth muscle contraction
G-protein coupled acetylcholine receptor signaling pathway
digestion
regulation of vasoconstriction
positive regulation of smooth muscle contraction
saliva secretion
protein binding
drug binding
G-protein coupled acetylcholine receptor activity
acetylcholine binding
regulation of vascular smooth muscle contraction
smooth muscle contraction
G-protein coupled acetylcholine receptor signaling pathway
digestion
regulation of vasoconstriction
positive regulation of smooth muscle contraction
saliva secretion
protein binding
drug binding
G-protein coupled acetylcholine receptor activity
acetylcholine binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3955 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036509 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036509 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036509 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P455531 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1331 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73418 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55784 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65894 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7819 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30768 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93888 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58528 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17196 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29824 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82810 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75866 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104777 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10795903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795899 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10795898 | - | - |
| Rat Genome 230 2.0 Array | 1369112_at | - | - |
| Rat Genome 230 2.0 Array | 1369112_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387462_at | - | - |
| Rat Genome 230 2.0 Array | 1387462_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387462_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387462_at | - | - |
| Affymetrix HT Rat Focus Array | 1369112_at | - | - |
| Affymetrix HT Rat Focus Array | 1369112_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700013730.rn.1 | TC1700002022.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387462_at | - | - |
| Rat Expression Set 230 | 1369112_at | - | - |
| Rat Expression Set 230 | 1369112_PM_at | - | - |
| Rat Expression Set 230 | 1387462_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795898 | - | - |
| Rat Genome U34 Set | AB017656_s_at | - | - |
| Rat Genome U34 Set | M16407_at | - | - |
| Rat Genome U34 Set | M16407_at | - | - |
| Rat Genome U34 Set | AB017656_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20717022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351883 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62650 | - | - |

Chrm3
Genomic Map