Assay Details
| Gene Symbol: Npy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012614.2 | NP_036746.1 | 1 - 2 | 96 | 63 | |
| GenBank mRNA | M15880.1 | - | 1 - 2 | 71 | 63 | |
| M20373.1 | - | 1 - 2 | 51 | 63 | ||
| Gene Symbol: LOC100912228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749745.3 | XP_003749793.1 | 109 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the M15880.1,M20373.1,NM_012614.2,XM_003749745.3 transcripts.
This assay is in the 5'UTR region of the M15880.1,M20373.1,NM_012614.2,XM_003749745.3 transcripts.
Set Membership: |
|

Npy
Genomic Map