Assay Details
| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005246.1 | NP_001005246.1 | 3 - 4 | 247 | 75 | |
| XM_008773252.1 | XP_008771474.1 | 3 - 4 | 314 | 75 | ||
| XM_008773253.1 | XP_008771475.1 | 3 - 4 | 314 | 75 | ||
| GenBank mRNA | X65468.1 | - | 3 - 4 | 262 | 75 | |
| X69767.1 | - | 3 - 4 | 262 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-matrix adhesion
regulation of heart rate
regulation of transcription, DNA-templated
muscle organ development
skeletal muscle tissue development
aging
establishment of blood-nerve barrier
regulation of gene expression
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of neuron projection development
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
myotube cell development
olfactory nerve structural organization
cell differentiation
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
regulation of membrane potential
muscle cell differentiation
peptide biosynthetic process
cellular protein complex assembly
neurotransmitter receptor metabolic process
positive regulation of neuron differentiation
muscle cell cellular homeostasis
muscle fiber development
neuron projection morphogenesis
nucleus localization
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
establishment of glial blood-brain barrier
negative regulation of ERK1 and ERK2 cascade
cardiac muscle cell action potential
regulation of cellular response to growth factor stimulus
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
integrin binding
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
PDZ domain binding
protein complex binding
nitric-oxide synthase binding
positive regulation of cell-matrix adhesion
regulation of heart rate
regulation of transcription, DNA-templated
muscle organ development
skeletal muscle tissue development
aging
establishment of blood-nerve barrier
regulation of gene expression
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of neuron projection development
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
myotube cell development
olfactory nerve structural organization
cell differentiation
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
regulation of membrane potential
muscle cell differentiation
peptide biosynthetic process
cellular protein complex assembly
neurotransmitter receptor metabolic process
positive regulation of neuron differentiation
muscle cell cellular homeostasis
muscle fiber development
neuron projection morphogenesis
nucleus localization
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
establishment of glial blood-brain barrier
negative regulation of ERK1 and ERK2 cascade
cardiac muscle cell action potential
regulation of cellular response to growth factor stimulus
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
integrin binding
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
PDZ domain binding
protein complex binding
nitric-oxide synthase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P303640 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P524536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18056 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P524536 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P303640 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P303640 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P524536 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P524536 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P303640 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P308877 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P353654 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36061 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47830 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92412 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4434 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78114 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99736 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72550 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62707 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52024 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4989 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26854 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10933882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933887 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933898 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933897 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933889 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10933819 | - | - |
| Rat Genome 230 2.0 Array | 1369810_at | - | - |
| Rat Genome 230 2.0 Array | 1369810_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369810_at | - | - |
| Affymetrix HT Rat Focus Array | 1369810_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00017160.rn.1 | TC0X00002685.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369810_PM_at | - | - |
| Rat Expression Set 230 | 1369810_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10933819 | - | - |
| Rat Genome U34 Set | X69767_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20898510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376680 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68602 | - | - |

Dmd
Genomic Map