Assay Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013136.1 | NP_037268.1 | 11 - 12 | 1823 | 64 | |
| GenBank mRNA | M35862.1 | - | 11 - 12 | 1823 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
multicellular organism development
spermatogenesis
cell differentiation
photoreceptor cell maintenance
protein autophosphorylation
regulation of cell cycle
transcription coactivator activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
multicellular organism development
spermatogenesis
cell differentiation
photoreceptor cell maintenance
protein autophosphorylation
regulation of cell cycle
transcription coactivator activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026122 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P157642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154415 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026122 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P157642 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026122 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P157642 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026122 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P157642 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146988 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99602 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84858 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10794635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794626 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794627 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794629 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794636 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10794625 | - | - |
| Rat Genome 230 2.0 Array | 1387324_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387324_at | - | - |
| Affymetrix HT Rat Focus Array | 1387324_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387324_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700002770.rn.1 | TC1700000364.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387324_at | - | - |
| Rat Expression Set 230 | 1387324_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10794625 | - | - |
| Rat Genome U34 Set | M35862_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20802675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373612 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63890 | - | - |

Mak
Genomic Map