Assay Details
| Gene Symbol: Hcrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013179.2 | NP_037311.1 | 1 - 2 | 108 | 148 | |
| GenBank mRNA | AF019565.1 | - | 1 - 2 | 113 | 148 | |
| AF041241.1 | - | 1 - 2 | 112 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF019565.1, AF041241.1, NM_013179.2 transcripts.
Set Membership: |
|

Hcrt
Genomic Map