Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013217.2 | NP_037349.1 | 30 - 31 | 5287 | 61 | |
| GenBank mRNA | CA511204.1 | - | 2 - 3 | 446 | 61 | |
| U83230.1 | - | 30 - 31 | 5287 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P165222 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P381197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58677 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165222 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P794669 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165222 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P794669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P794669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P165222 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P239985 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P287911 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701491469 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67083 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68581 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49414 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36200 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59283 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82860 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701491469 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10703214 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703205 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703196 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703197 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703212 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703213 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703210 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703198 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703208 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703209 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703195 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886000 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10703189 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10885998 | - | - |
| Rat Genome 230 2.0 Array | 1387392_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388036_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1373027_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373027_at | - | - |
| Rat Genome 230 2.0 Array | 1388036_a_at | - | - |
| Rat Genome 230 2.0 Array | 1387392_at | - | - |
| Affymetrix HT Rat Focus Array | 1387392_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373027_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388036_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1373027_at | - | - |
| Affymetrix HT Rat Focus Array | 1387392_at | - | - |
| Affymetrix HT Rat Focus Array | 1388036_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100035819.rn.1 | TC0100004521.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100035818.rn.1 | TC0100004521.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373027_PM_at | - | - |
| Rat Expression Set 230 | 1388036_PM_a_at | - | - |
| Rat Expression Set 230 | 1388036_a_at | - | - |
| Rat Expression Set 230 | 1387392_PM_at | - | - |
| Rat Expression Set 230 | 1373027_at | - | - |
| Rat Expression Set 230 | 1387392_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10703189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885998 | - | - |
| Rat Genome U34 Set | U83230_at | - | - |
| Rat Genome U34 Set | rc_AA859702_at | - | - |
| Rat Genome U34 Set | rc_AI071511_s_at | - | - |
| Rat Genome U34 Set | rc_AI170067_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21200480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359100 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66066 | - | - |

Mllt4
Genomic Map