Assay Details
| Gene Symbol: Kpnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017063.1 | NP_058759.1 | 15 - 16 | 2014 | 75 | |
| GenBank mRNA | L38644.1 | - | 15 - 16 | 2014 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P543350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79257 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P543350 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P543350 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P543350 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P543350 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700180904 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55355 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81191 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700180904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10746671 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746662 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746667 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746665 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746669 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746673 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746672 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746666 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746660 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746663 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746664 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746657 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746661 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746656 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746655 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746658 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746659 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746668 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746670 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746674 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746675 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746676 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10746654 | - | - |
| Rat Genome 230 2.0 Array | 1372550_at | - | - |
| Rat Genome 230 2.0 Array | 1372550_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368573_at | - | - |
| Rat Genome 230 2.0 Array | 1368573_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368573_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368573_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000032790.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032793.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032797.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032799.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032807.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032808.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032796.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032792.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032809.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032811.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032802.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032810.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032812.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032814.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032817.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032819.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032805.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032794.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032804.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032800.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032803.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032806.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032813.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032815.rn.1 | TC1000003679.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032816.rn.1 | TC1000003679.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368573_PM_at | - | - |
| Rat Expression Set 230 | 1368573_at | - | - |
| Rat Expression Set 230 | 1372550_PM_at | - | - |
| Rat Expression Set 230 | 1372550_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746654 | - | - |
| Rat Genome U34 Set | L38644_at | - | - |
| Rat Genome U34 Set | rc_AA851873_at | - | - |
| Rat Genome U34 Set | rc_AI169278_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22359378 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373492 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69975 | - | - |

Kpnb1
Genomic Map