Assay Details
| Gene Symbol: Kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017099.4 | NP_058795.3 | 1 - 2 | 117 | 109 | |
| GenBank mRNA | BC078863.1 | - | 100 | 7128284 | 109 | |
| D42145.1 | - | 1 - 2 | 120 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078863.1, D42145.1, NM_017099.4 transcripts.
Set Membership: |
|

Kcnj8
Genomic Map