Assay Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017202.1 | NP_058898.1 | 1 - 2 | 88 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_017202.1 transcripts.
Set Membership: |
|

Cox4i1
Genomic Map