Assay Details
| Gene Symbol: Erbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017218.3 | NP_058914.2 | 1 - 2 | 226 | 61 | |
| GenBank mRNA | U29339.2 | - | 1 - 2 | 226 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocardial cushion development
negative regulation of cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
axonogenesis
peripheral nervous system development
skeletal muscle tissue development
circadian rhythm
response to wounding
negative regulation of signal transduction
Schwann cell differentiation
phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cranial nerve development
signal transduction by protein phosphorylation
cellular response to insulin stimulus
regulation of cell proliferation
odontogenesis
response to drug
negative regulation of neuron apoptotic process
tongue development
positive regulation of glucose import
negative regulation of secretion
neuron apoptotic process
mammary gland involution
positive regulation of protein tyrosine kinase activity
positive regulation of calcineurin-NFAT signaling cascade
extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
ATP binding
growth factor binding
protein tyrosine kinase activator activity
neuregulin receptor activity
neuregulin binding
identical protein binding
protein heterodimerization activity
endocardial cushion development
negative regulation of cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
axonogenesis
peripheral nervous system development
skeletal muscle tissue development
circadian rhythm
response to wounding
negative regulation of signal transduction
Schwann cell differentiation
phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cranial nerve development
signal transduction by protein phosphorylation
cellular response to insulin stimulus
regulation of cell proliferation
odontogenesis
response to drug
negative regulation of neuron apoptotic process
tongue development
positive regulation of glucose import
negative regulation of secretion
neuron apoptotic process
mammary gland involution
positive regulation of protein tyrosine kinase activity
positive regulation of calcineurin-NFAT signaling cascade
extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
ATP binding
growth factor binding
protein tyrosine kinase activator activity
neuregulin receptor activity
neuregulin binding
identical protein binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P080643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161129 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080643 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P080643 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P080643 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P557246 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90547 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17787 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94920 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10899847 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899865 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899860 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899850 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899858 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899845 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899844 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899864 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899862 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899859 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899857 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899843 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899851 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899848 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899866 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899854 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899846 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899849 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899856 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899867 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899855 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899861 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899841 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10899839 | - | - |
| Rat Genome 230 2.0 Array | 1369088_at | - | - |
| Rat Genome 230 2.0 Array | 1369088_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377821_at | - | - |
| Rat Genome 230 2.0 Array | 1377821_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369088_at | - | - |
| Affymetrix HT Rat Focus Array | 1369088_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700015704.rn.1 | TC0700002000.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700015701.rn.1 | TC0700002000.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369088_at | - | - |
| Rat Expression Set 230 | 1369088_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10899839 | - | - |
| Rat Genome U34 Set | U29339_at | - | - |
| Rat Genome U34 Set | U52530_s_at | - | - |
| Rat Genome U34 Set | rc_AA924236_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21201447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353005 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59660 | - | - |

Erbb3
Genomic Map