Assay Details
| Gene Symbol: Cdon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017358.1 | NP_059054.1 | 16 - 17 | 3302 | 60 | |
| XM_006242791.2 | XP_006242853.1 | 16 - 17 | 3340 | 60 | ||
| XM_006242792.2 | XP_006242854.1 | 17 - 18 | 3929 | 60 | ||
| XM_006242793.2 | XP_006242855.1 | 15 - 16 | 2986 | 60 | ||
| XM_008766066.1 | XP_008764288.1 | 17 - 18 | 4739 | 60 | ||
| GenBank mRNA | AF004840.1 | - | 16 - 17 | 3302 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cell fate specification
positive regulation of protein phosphorylation
lens development in camera-type eye
cell adhesion
smoothened signaling pathway
myoblast fusion
anterior/posterior pattern specification
embryonic body morphogenesis
skeletal muscle satellite cell differentiation
cerebral cortex development
positive regulation of MAPK cascade
regulation of protein heterodimerization activity
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic morphogenesis
positive regulation of skeletal muscle tissue development
positive regulation of small GTPase mediated signal transduction
striated muscle cell differentiation
embryonic retina morphogenesis in camera-type eye
positive regulation of neural precursor cell proliferation
molecular_function
protein binding
cell fate specification
positive regulation of protein phosphorylation
lens development in camera-type eye
cell adhesion
smoothened signaling pathway
myoblast fusion
anterior/posterior pattern specification
embryonic body morphogenesis
skeletal muscle satellite cell differentiation
cerebral cortex development
positive regulation of MAPK cascade
regulation of protein heterodimerization activity
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic morphogenesis
positive regulation of skeletal muscle tissue development
positive regulation of small GTPase mediated signal transduction
striated muscle cell differentiation
embryonic retina morphogenesis in camera-type eye
positive regulation of neural precursor cell proliferation
molecular_function
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P011419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176716 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P011419 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P011419 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P011419 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11935 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47236 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41645 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91433 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10908970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908969 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908978 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10908968 | - | - |
| Rat Genome 230 2.0 Array | 1369016_at | - | - |
| Rat Genome 230 2.0 Array | 1369016_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369016_at | - | - |
| Affymetrix HT Rat Focus Array | 1369016_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800002505.rn.1 | TC0800000373.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369016_at | - | - |
| Rat Expression Set 230 | 1369016_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908968 | - | - |
| Rat Genome U34 Set | AF004840_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21981448 | - | - |

Cdon
Genomic Map