Assay Details
| Gene Symbol: Gjb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019240.1 | NP_062113.1 | 1 | 652 | 66 | |
| XM_006238845.2 | XP_006238907.1 | 2 | 1962 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gjb3
Genomic Map