Assay Details
| Gene Symbol: Pitx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019247.1 | NP_062120.1 | 1 - 2 | 57 | 60 | |
| GenBank mRNA | AJ011005.2 | - | 1 - 2 | 57 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ011005.2, NM_019247.1 transcripts.
Set Membership: |
|

Pitx3
Genomic Map