Assay Details
| Gene Symbol: Ninj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021595.2 | NP_067606.1 | 1 - 2 | 168 | 75 | |
| GenBank mRNA | AB040815.1 | - | 1 - 2 | 169 | 75 | |
| AF250322.1 | - | 1 - 2 | 169 | 75 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P384260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18442 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P384260 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P384260 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P384260 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P384260 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700248377 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33813 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76106 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4528 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40401 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10129 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21610 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99682 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700248377 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10858280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858281 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10858277 | - | - |
| Rat Genome 230 2.0 Array | 1368135_at | - | - |
| Rat Genome 230 2.0 Array | 1368135_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368135_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368135_at | - | - |
| Rat Expression Set 230 | 1368135_at | - | - |
| Rat Expression Set 230 | 1368135_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20797910 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370725 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62813 | - | - |

Ninj2
Genomic Map