Assay Details
| Gene Symbol: Kcnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021688.3 | NP_067720.1 | 1 - 2 | 374 | 111 | |
| GenBank mRNA | AF022819.1 | - | 1 - 2 | 396 | 111 | |
| BC061807.1 | - | 1 - 2 | 374 | 5598415 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31939 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036890 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036890 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036890 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P102628 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700252269 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8666 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35945 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700252269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10808926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10808923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10808924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10808925 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10808922 | - | - |
| Rat Genome 230 2.0 Array | 1369500_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369500_at | - | - |
| Affymetrix HT Rat Focus Array | 1369500_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369500_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900007072.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007073.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007075.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007086.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007082.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007087.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007076.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007085.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007074.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007077.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007089.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007078.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007083.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007088.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007090.rn.1 | TC1900000871.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900007081.rn.1 | TC1900000871.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369500_at | - | - |
| Rat Expression Set 230 | 1369500_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10808922 | - | - |
| Rat Genome U34 Set | AF022819_at | - | - |
| Rat Genome U34 Set | rc_AA926247_at | - | - |
| Rat Genome U34 Set | AF022819_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21106682 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356313 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53977 | - | - |

Kcnk1
Genomic Map