Assay Details
| Gene Symbol: Exoc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022204.3 | NP_071540.1 | 3 - 4 | 504 | 93 | |
| GenBank mRNA | BC128739.1 | - | 3 - 4 | 507 | 8362342 | 93 |
| U79417.1 | - | 3 - 4 | 276 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P188596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P188596 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P188596 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P188596 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P188596 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29577 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12969 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69848 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10782991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783001 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10782990 | - | - |
| Rat Genome 230 2.0 Array | 1395155_at | - | - |
| Rat Genome 230 2.0 Array | 1387841_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387841_at | - | - |
| Rat Genome 230 2.0 Array | 1395155_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387841_at | - | - |
| Affymetrix HT Rat Focus Array | 1387841_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395155_at | - | - |
| Affymetrix HT Rat Focus Array | 1395155_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500009649.rn.1 | TC1500001526.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387841_at | - | - |
| Rat Expression Set 230 | 1387841_PM_at | - | - |
| Rat Expression Set 230 | 1395155_at | - | - |
| Rat Expression Set 230 | 1395155_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10782990 | - | - |
| Rat Genome U34 Set | U79417_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21230568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353046 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52081 | - | - |

Exoc5
Genomic Map