Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022246.1 | NP_071582.1 | 13 - 14 | 2364 | 103 | |
| GenBank mRNA | AF218576.1 | - | 13 - 14 | 2364 | 103 | |
| FQ232550.1 | - | 11 - 12 | 1888 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mitotic recombination
telomere maintenance
DNA catabolic process, endonucleolytic
DNA repair
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
telomere maintenance via telomerase
reciprocal meiotic recombination
regulation of mitotic cell cycle
positive regulation of protein autophosphorylation
DNA duplex unwinding
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
protein heterooligomerization
chromosome organization involved in meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
3'-5' exonuclease activity
ATPase activity
protein binding, bridging
metal ion binding
regulation of mitotic recombination
telomere maintenance
DNA catabolic process, endonucleolytic
DNA repair
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
telomere maintenance via telomerase
reciprocal meiotic recombination
regulation of mitotic cell cycle
positive regulation of protein autophosphorylation
DNA duplex unwinding
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
protein heterooligomerization
chromosome organization involved in meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
DNA binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
3'-5' exonuclease activity
ATPase activity
protein binding, bridging
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P169307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27940 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P169307 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P169307 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P169307 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P169307 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700775240 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700885159 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702155843 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71305 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1039 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57314 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98965 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700885159 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700775240 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702155843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10742623 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742625 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742629 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742624 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742620 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742626 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742627 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10742617 | - | - |
| Rat Genome 230 2.0 Array | 1397642_at | - | - |
| Rat Genome 230 2.0 Array | 1387427_at | - | - |
| Rat Genome 230 2.0 Array | 1385565_at | - | - |
| Rat Genome 230 2.0 Array | 1397642_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385565_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387427_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397642_at | - | - |
| Affymetrix HT Rat Focus Array | 1385565_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385565_at | - | - |
| Affymetrix HT Rat Focus Array | 1387427_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397642_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387427_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000024502.rn.1 | TC1000002793.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387427_at | - | - |
| Rat Expression Set 230 | 1387427_PM_at | - | - |
| Rat Expression Set 230 | 1397642_at | - | - |
| Rat Expression Set 230 | 1385565_PM_at | - | - |
| Rat Expression Set 230 | 1397642_PM_at | - | - |
| Rat Expression Set 230 | 1385565_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21912076 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354275 | - | - |
| RatRef-12 Expression BeadChip | ILMN_61480 | - | - |

Rad50
Genomic Map