Assay Details
| Gene Symbol: Tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022589.1 | NP_072111.1 | 2 - 3 | 190 | 58 | |
| GenBank mRNA | AJ271442.1 | - | 2 - 3 | 190 | 58 | |
| BC085733.1 | - | 2 - 3 | 210 | 7122828 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P005243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107827 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P005243 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P005243 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P005243 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P692798 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P344444 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702131139 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66157 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97196 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31372 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702131139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10817907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817905 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817904 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10817898 | - | - |
| Rat Genome 230 2.0 Array | 1368105_at | - | - |
| Rat Genome 230 2.0 Array | 1368104_at | - | - |
| Rat Genome 230 2.0 Array | 1368105_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368104_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368105_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368105_at | - | - |
| Affymetrix HT Rat Focus Array | 1368104_at | - | - |
| Affymetrix HT Rat Focus Array | 1368104_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200012683.rn.1 | TC0200001870.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368105_at | - | - |
| Rat Expression Set 230 | 1368104_PM_at | - | - |
| Rat Expression Set 230 | 1368105_PM_at | - | - |
| Rat Expression Set 230 | 1368104_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10817898 | - | - |
| Rat Genome U34 Set | rc_AA899764_at | - | - |
| Rat Genome U34 Set | rc_AI228231_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21201924 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370492 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60033 | - | - |

Tspan2
Genomic Map