Assay Details
| Gene Symbol: Fat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022954.1 | NP_075243.1 | 19 - 20 | 11598 | 83 | |
| GenBank mRNA | AB011527.1 | - | 19 - 20 | 11598 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P065600 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P550241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69939 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065600 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P550241 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P550241 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P065600 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P550241 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P065600 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P550241 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21606 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64325 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48250 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1932 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103822 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10742800 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742787 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742780 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742799 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742801 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742798 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742779 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742795 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742797 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742778 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742782 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10742777 | - | - |
| Rat Genome 230 2.0 Array | 1368684_at | - | - |
| Rat Genome 230 2.0 Array | 1368684_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368684_at | - | - |
| Affymetrix HT Rat Focus Array | 1368684_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000024808.rn.1 | TC1000002824.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368684_PM_at | - | - |
| Rat Expression Set 230 | 1368684_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742777 | - | - |
| Rat Genome U34 Set | AB011527_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20909262 | - | - |

Fat2
Genomic Map