Assay Details
| Gene Symbol: Mtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030864.1 | NP_110491.1 | 27 - 28 | 3101 | 61 | |
| GenBank mRNA | AF034214.1 | - | 27 - 28 | 3101 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein methylation
methionine metabolic process
methionine biosynthetic process
methylation
pteridine-containing compound metabolic process
tetrahydrofolate metabolic process
homocysteine metabolic process
cellular response to methionine
folic acid binding
methyltransferase activity
zinc ion binding
methionine synthase activity
S-adenosylmethionine-homocysteine S-methyltransferase activity
amino acid binding
cobalamin binding
protein methylation
methionine metabolic process
methionine biosynthetic process
methylation
pteridine-containing compound metabolic process
tetrahydrofolate metabolic process
homocysteine metabolic process
cellular response to methionine
folic acid binding
methyltransferase activity
zinc ion binding
methionine synthase activity
S-adenosylmethionine-homocysteine S-methyltransferase activity
amino acid binding
cobalamin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P479917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99822 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P479917 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P479917 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P479917 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P479917 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14118 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15286 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28144 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53394 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 392 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10795790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795777 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795778 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795775 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795772 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795767 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795780 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795769 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795773 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795779 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795762 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795764 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795761 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795768 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795774 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795771 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795765 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795776 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795770 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795766 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795763 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795787 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10795760 | - | - |
| Rat Genome 230 2.0 Array | 1387545_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387545_at | - | - |
| Affymetrix HT Rat Focus Array | 1387545_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387545_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700013894.rn.1 | TC1700002036.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387545_at | - | - |
| Rat Expression Set 230 | 1387545_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795760 | - | - |
| Rat Genome U34 Set | AF034214_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21563506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373194 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69292 | - | - |

Mtr
Genomic Map