Assay Details
| Gene Symbol: Cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031241.1 | NP_112520.1 | 1 | 948 | 93 | |
| GenBank mRNA | AB009686.1 | - | 1 | 948 | 93 | |
| AB018596.1 | - | 1 | 915 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cyp8b1
Genomic Map