Assay Details
| Gene Symbol: Mkln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031359.1 | NP_112649.1 | 1 - 2 | 102 | 73 | |
| GenBank mRNA | AB046442.1 | - | 1 - 2 | 102 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1040601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19287 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1040601 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1040601 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1040601 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1040601 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43653 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82386 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20250 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57337 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20122 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86813 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30851 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78237 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65390 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10854356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854341 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854355 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10854338 | - | - |
| Rat Genome 230 2.0 Array | 1368018_at | - | - |
| Rat Genome 230 2.0 Array | 1368018_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368018_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368018_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400004120.rn.1 | TC0400000577.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400004121.rn.1 | TC0400000577.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368018_PM_at | - | - |
| Rat Expression Set 230 | 1368018_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10854338 | - | - |
| Rat Genome U34 Set | rc_AI008772_at | - | - |
| Rat Genome U34 Set | rc_AA946519_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20910169 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351935 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69118 | - | - |

Mkln1
Genomic Map