Assay Details
| Gene Symbol: Rab2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031718.1 | NP_113906.1 | 3 - 4 | 185 | 114 | |
| GenBank mRNA | FQ229541.1 | - | 3 - 4 | 387 | 114 | |
| J02999.1 | - | 3 - 4 | 185 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P390086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85400 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P390086 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P390086 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P390086 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P390086 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73675 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46827 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11504 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33310 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20374 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35861 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10867536 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867532 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867533 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867535 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867534 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867531 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10867528 | - | - |
| Rat Genome 230 2.0 Array | 1370087_at | - | - |
| Rat Genome 230 2.0 Array | 1392681_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370087_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392681_at | - | - |
| Affymetrix HT Rat Focus Array | 1370087_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370087_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500001059.rn.1 | TC0500000165.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500001063.rn.1 | TC0500000165.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500001062.rn.1 | TC0500000165.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500001061.rn.1 | TC0500000165.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370087_at | - | - |
| Rat Expression Set 230 | 1370087_PM_at | - | - |
| Rat Expression Set 230 | 1392681_PM_at | - | - |
| Rat Expression Set 230 | 1392681_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10867528 | - | - |
| Rat Genome U34 Set | rc_AI009095_at | - | - |
| Rat Genome U34 Set | rc_AI237381_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21043828 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369180 | - | - |
| RatRef-12 Expression BeadChip | ILMN_61489 | - | - |

Rab2a
Genomic Map