Assay Details
| Gene Symbol: Hspb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031750.1 | NP_113938.1 | 1 | 456 | 61 | |
| GenBank mRNA | AF203374.1 | - | 1 | 456 | 61 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hspb3
Genomic Map