Assay Details
| Gene Symbol: Insl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053680.1 | NP_446132.1 | 1 - 2 | 174 | 90 | |
| GenBank mRNA | AF139918.2 | - | 1 - 2 | 174 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Insl3
Genomic Map