Assay Details
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053840.2 | NP_446292.2 | 582 | 91 | ||
| GenBank mRNA | BC078768.1 | - | 582 | 7133071 | 91 | |
| M33821.1 | - | 680 | 92 | |||
| M33822.1 | - | 667 | 91 | |||
| X15443.1 | - | 588 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ggt1
Genomic Map