Assay Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080906.2 | NP_543182.1 | 1 - 2 | 122 | 68 | |
| GenBank mRNA | AF334162.1 | - | 1 - 2 | 127 | 68 | |
| BC062021.1 | - | 1 - 2 | 55 | 5623446 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF334162.1, BC062021.1, NM_080906.2 transcripts.
Set Membership: |
|

Ddit4
Genomic Map