Assay Details
| Gene Symbol: Ep300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765878.1 | XP_008764100.1 | 1 - 2 | 1065 | 129 | |
| GenBank mRNA | AB066220.1 | - | 1 - 2 | 95 | 129 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
somitogenesis
liver development
positive regulation of protein phosphorylation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein acetylation
internal protein amino acid acetylation
apoptotic process
positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
heart development
skeletal muscle tissue development
memory
circadian rhythm
response to glucose
organ morphogenesis
positive regulation of glycoprotein biosynthetic process
positive regulation of gene expression
positive regulation of cell death
response to organic cyclic compound
positive regulation of muscle atrophy
histone acetylation
N-terminal peptidyl-lysine acetylation
internal peptidyl-lysine acetylation
positive regulation of cell growth
lung development
negative regulation of cellular metabolic process
positive regulation of cellular metabolic process
response to cobalt ion
positive regulation of protein binding
response to retinoic acid
positive regulation of collagen biosynthetic process
positive regulation of protein import into nucleus, translocation
response to tumor necrosis factor
positive regulation of histone acetylation
cellular response to drug
cellular response to trichostatin A
response to drug
response to hydrogen peroxide
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA binding
protein kinase B signaling
response to estrogen
positive regulation by host of viral transcription
histone H3 acetylation
histone H4 acetylation
histone H2B acetylation
response to ethanol
positive regulation of translation
positive regulation of axon extension
positive regulation of cell size
positive regulation of proteolysis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
positive regulation of protein secretion
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
response to calcium ion
regulation of angiotensin metabolic process
positive regulation of sarcomere organization
negative regulation of cell death
regulation of androgen receptor signaling pathway
protein-DNA complex assembly
cellular response to hydrogen peroxide
response to fatty acid
cellular response to antibiotic
cellular response to retinoic acid
cellular response to cAMP
cellular response to glucose stimulus
cellular response to mineralocorticoid stimulus
cellular response to organic cyclic compound
response to dexamethasone
cellular response to dexamethasone stimulus
regulation of tubulin deacetylation
positive regulation of protein acetylation
regulation of nucleic acid-templated transcription
cellular response to nerve growth factor stimulus
negative regulation of miRNA metabolic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
core promoter proximal region DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
p53 binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
antigen binding
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
protein binding
beta-catenin binding
transcription factor binding
acetyltransferase activity
transferase activity, transferring acyl groups
protein kinase binding
chromatin DNA binding
protein complex binding
activating transcription factor binding
nuclear hormone receptor binding
glucocorticoid receptor binding
peroxisome proliferator activated receptor binding
bHLH transcription factor binding
SMAD binding
androgen receptor binding
mitogen-activated protein kinase binding
NF-kappaB binding
pre-mRNA intronic binding
protein antigen binding
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
somitogenesis
liver development
positive regulation of protein phosphorylation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein acetylation
internal protein amino acid acetylation
apoptotic process
positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
heart development
skeletal muscle tissue development
memory
circadian rhythm
response to glucose
organ morphogenesis
positive regulation of glycoprotein biosynthetic process
positive regulation of gene expression
positive regulation of cell death
response to organic cyclic compound
positive regulation of muscle atrophy
histone acetylation
N-terminal peptidyl-lysine acetylation
internal peptidyl-lysine acetylation
positive regulation of cell growth
lung development
negative regulation of cellular metabolic process
positive regulation of cellular metabolic process
response to cobalt ion
positive regulation of protein binding
response to retinoic acid
positive regulation of collagen biosynthetic process
positive regulation of protein import into nucleus, translocation
response to tumor necrosis factor
positive regulation of histone acetylation
cellular response to drug
cellular response to trichostatin A
response to drug
response to hydrogen peroxide
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA binding
protein kinase B signaling
response to estrogen
positive regulation by host of viral transcription
histone H3 acetylation
histone H4 acetylation
histone H2B acetylation
response to ethanol
positive regulation of translation
positive regulation of axon extension
positive regulation of cell size
positive regulation of proteolysis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
positive regulation of protein secretion
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
response to calcium ion
regulation of angiotensin metabolic process
positive regulation of sarcomere organization
negative regulation of cell death
regulation of androgen receptor signaling pathway
protein-DNA complex assembly
cellular response to hydrogen peroxide
response to fatty acid
cellular response to antibiotic
cellular response to retinoic acid
cellular response to cAMP
cellular response to glucose stimulus
cellular response to mineralocorticoid stimulus
cellular response to organic cyclic compound
response to dexamethasone
cellular response to dexamethasone stimulus
regulation of tubulin deacetylation
positive regulation of protein acetylation
regulation of nucleic acid-templated transcription
cellular response to nerve growth factor stimulus
negative regulation of miRNA metabolic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
core promoter proximal region DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
p53 binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
antigen binding
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
protein binding
beta-catenin binding
transcription factor binding
acetyltransferase activity
transferase activity, transferring acyl groups
protein kinase binding
chromatin DNA binding
protein complex binding
activating transcription factor binding
nuclear hormone receptor binding
glucocorticoid receptor binding
peroxisome proliferator activated receptor binding
bHLH transcription factor binding
SMAD binding
androgen receptor binding
mitogen-activated protein kinase binding
NF-kappaB binding
pre-mRNA intronic binding
protein antigen binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P822616 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P822616 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P822616 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P822616 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15436 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P822616 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68340 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26191 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10897923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897920 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10897919 | - | - |
| Rat Genome 230 2.0 Array | 1373916_at | - | - |
| Rat Genome 230 2.0 Array | 1369307_at | - | - |
| Rat Genome 230 2.0 Array | 1369307_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373916_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369307_at | - | - |
| Affymetrix HT Rat Focus Array | 1373916_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373916_at | - | - |
| Affymetrix HT Rat Focus Array | 1369307_PM_at | - | - |
| Rat Expression Set 230 | 1373916_at | - | - |
| Rat Expression Set 230 | 1369307_at | - | - |
| Rat Expression Set 230 | 1369307_PM_at | - | - |
| Rat Expression Set 230 | 1373916_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10897919 | - | - |
| Rat Genome U34 Set | rc_AA996888_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22381552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2039699 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64190 | - | - |

Ep300
Genomic Map