Assay Details
| Gene Symbol: Uggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133596.1 | NP_598280.1 | 25 - 26 | 2796 | 65 | |
| XM_006244734.2 | XP_006244796.1 | 25 - 26 | 3018 | 65 | ||
| XM_006244735.2 | XP_006244797.1 | 24 - 25 | 2994 | 65 | ||
| GenBank mRNA | AF200359.1 | - | 25 - 26 | 2796 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P550203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115333 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P550203 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P550203 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P550203 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P550203 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701257948 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4921 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84408 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42736 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73519 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50113 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68129 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17453 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701257948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10927295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927315 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927314 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927312 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927320 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927311 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927308 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927310 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927313 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927316 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927317 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927318 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927297 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10927285 | - | - |
| Rat Genome 230 2.0 Array | 1368953_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368953_at | - | - |
| Affymetrix HT Rat Focus Array | 1368953_at | - | - |
| Affymetrix HT Rat Focus Array | 1368953_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900014720.rn.1 | TC0900001809.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368953_PM_at | - | - |
| Rat Expression Set 230 | 1368953_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10927285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22168465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364029 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62615 | - | - |

Uggt1
Genomic Map