Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139091.2 | NP_620791.1 | 1 - 2 | 190 | 77 | |
| XM_006252093.2 | XP_006252155.1 | 1 - 2 | 295 | 77 | ||
| GenBank mRNA | AF000900.1 | - | 1 - 2 | 127 | 77 | |
| BC085690.1 | - | 1 - 2 | 190 | 7190650 | 77 | |
| U63839.1 | - | 1 - 2 | 203 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117731 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047512 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74767 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047512 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117731 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117731 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047507 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047512 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117731 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047507 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047512 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231283 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P318607 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P524578 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P524576 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71711 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35735 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66009 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784329 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784327 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784313 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784310 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784315 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784325 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784317 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784320 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784318 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784316 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784314 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784328 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784312 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784307 | - | - |
| Rat Genome 230 2.0 Array | 1388073_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370437_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371207_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388073_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1371208_at | - | - |
| Rat Genome 230 2.0 Array | 1370437_at | - | - |
| Rat Genome 230 2.0 Array | 1388198_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388198_at | - | - |
| Rat Genome 230 2.0 Array | 1371207_at | - | - |
| Rat Genome 230 2.0 Array | 1371208_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371207_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370437_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388073_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371208_at | - | - |
| Affymetrix HT Rat Focus Array | 1388198_at | - | - |
| Affymetrix HT Rat Focus Array | 1371207_at | - | - |
| Affymetrix HT Rat Focus Array | 1388198_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388073_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370437_at | - | - |
| Affymetrix HT Rat Focus Array | 1371208_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500012225.rn.1 | TC1500001814.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500012226.rn.1 | TC1500001814.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370437_at | - | - |
| Rat Expression Set 230 | 1388198_at | - | - |
| Rat Expression Set 230 | 1371208_at | - | - |
| Rat Expression Set 230 | 1371207_PM_at | - | - |
| Rat Expression Set 230 | 1371207_at | - | - |
| Rat Expression Set 230 | 1388198_PM_at | - | - |
| Rat Expression Set 230 | 1388073_PM_a_at | - | - |
| Rat Expression Set 230 | 1370437_PM_at | - | - |
| Rat Expression Set 230 | 1388073_a_at | - | - |
| Rat Expression Set 230 | 1371208_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784307 | - | - |
| Rat Genome U34 Set | U63839_at | - | - |
| Rat Genome U34 Set | AF000898_at | - | - |
| Rat Genome U34 Set | rc_AA891045_at | - | - |
| Rat Genome U34 Set | AF000899_g_at | - | - |
| Rat Genome U34 Set | AF000899_at | - | - |
| Rat Genome U34 Set | AF000901_s_at | - | - |
| Rat Genome U34 Set | rc_AI071478_i_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22004393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367491 | - | - |
| RatRef-12 Expression BeadChip | ILMN_57893 | - | - |

Nupl1
Genomic Map